Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetyldiphenyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. A235525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A235525G 25 g
A23555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A235525G Supplier TCI America Supplier No. A235525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10169-55-8
Molecular Formula C14H12OS
Molecular Weight (g/mol) 228.31
MDL Number MFCD00026227
InChI Key XUDYHODVSUXRPW-UHFFFAOYSA-N
Synonym 4-acetyldiphenyl sulfide, 1-4-phenylthio phenyl ethanone, 4-acetyl diphenyl sulfide, 4'-phenylthio acetophenone, 1-4-phenylsulfanyl phenyl ethanone, 1-4-phenylsulfanylphenyl ethanone, 1-4-phenylthio phenyl ethan-1-one, ethanone, 1-4-phenylthio phenyl, 1-acetyl-4-phenylthiobenzene
PubChem CID 66287
IUPAC Name 1-[4-(phenylsulfanyl)phenyl]ethan-1-one
SMILES CC(=O)C1=CC=C(SC2=CC=CC=C2)C=C1

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 184°C
Quantity 25 g
Formula Weight 228.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetyldiphenyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.