Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylphenyl isothiocyanate, 98%, Thermo Scientific™

Catalog No. AAL1192703
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-57-9
Molecular Formula C9H7NOS
Molecular Weight (g/mol) 177.221
MDL Number MFCD00041100
InChI Key KAKXOESNHIUYJT-UHFFFAOYSA-N
Synonym 4-acetylphenyl isothiocyanate, 1-4-isothiocyanatophenyl ethanone, 4-acetylphenylisothiocyanate, 1-4-isothiocyanatophenyl ethan-1-one, acmc-20ampw, bromobenzyl-4-isothiocyanate, 4'-isothiocyanatoacetophenone, 4-acetylphenyl-isothiocyanate, phenylisothiocyanate, 4-acetyl, 1-4-isothiocyanato-phenyl-ethanone
PubChem CID 242779
IUPAC Name 1-(4-isothiocyanatophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 77°C to 81°C
Boiling Point 112°C (6 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2207978
Sensitivity Moisture sensitive
Formula Weight 177.23
Percent Purity 98%
Chemical Name or Material 4-Acetylphenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.