Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acryloylmorpholine, 97%

Catalog No. ac439250500
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle

CAS: 5117-12-4 | C7H11NO2 | 141.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5117-12-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
InChI Key XLPJNCYCZORXHG-UHFFFAOYSA-N
SMILES C=CC(=O)N1CCOCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Yellow
Density 1.1220g/mL
Boiling Point 158.0°C (50.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 50 mL
Specific Gravity 1.122
Synonym 4-acryloylmorpholine, 1-morpholinoprop-2-en-1-one, n-acrylylmorpholine, n-acryloylmorpholine, morpholine, 4-1-oxo-2-propenyl, poly acryloylmorpholine, acryloylmorpholine, acryloyl morpholine, 1-morpholin-4-yl prop-2-en-1-one, unii-k0y58p61ja
IUPAC Name 1-morpholin-4-ylprop-2-en-1-one
PubChem CID 98723
Formula Weight 141.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Acryloylmorpholine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Get medical attention/advice if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.