Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Allylpyrocatechol 98.0+%, TCI America™
SDP

Catalog No. A2947100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2947100MG 100 mg
A294710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2947100MG Supplier TCI America Supplier No. A2947100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1126-61-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD01548379
InChI Key FHEHIXJLCWUPCZ-UHFFFAOYSA-N
Synonym 4-Allyl-1,2-benzenediol, 4-Allyl-1,2-dihydroxybenzene, 4-Allylcatechol, Hydroxychavicol
PubChem CID 70775
IUPAC Name 4-prop-2-enylbenzene-1,2-diol
SMILES C=CCC1=CC(=C(C=C1)O)O

Specifications

Melting Point 44°C
Boiling Point 123°C
Quantity 100 mg
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Allylpyrocatechol
RTECSNumber CZ9029440
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.