Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
4-Amino-1-benzylpiperidine, 98%, Thermo Scientific™
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100g
                            
                        
                    Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
 
            Specifications
Specifications
| CAS | 50541-93-0 | 
| Density | 0.93 | 
| Boiling Point | 279°C to 281°C | 
| Flash Point | >110°C (230°F) | 
| Molecular Formula | C12H18N2 | 
| Refractive Index | 1.543 | 
| Quantity | 100g | 
| MDL Number | MFCD00006504 | 
| Beilstein | 146038 | 
| Sensitivity | Air Sensitive | 
| Show More | 
Safety and Handling
| Hazard Category | H302+H312-H314-H335 | 
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. | 
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c | 
| DOTInformation | Hazard Class: 8; Packaging Group: II | 
| EINECSNumber | 256-620-9 | 
| TSCA | No | 
| Recommended Storage | Ambient temperatures; Store under Nitrogen | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            