Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-isopropylpiperidine 98.0+%, TCI America™
SDP

Catalog No. I077425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I077425G 25 g
I07745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I077425G Supplier TCI America Supplier No. I077425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127285-08-9
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.25
MDL Number MFCD03411606
InChI Key ZRQQXFMGYSOKDF-UHFFFAOYSA-N
PubChem CID 3163333
IUPAC Name 1-(propan-2-yl)piperidin-4-amine
SMILES CC(C)N1CCC(N)CC1

Specifications

Color Colorless
Quantity 25 g
UN Number 2920
Formula Weight 142.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amino-1-isopropylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.