Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass Bottle
14691-88-4
C9H19N2O
171.26
MFCD00006479
4-Amino-TEMPO
14691-88-4
C9H19N2O
171.26
MFCD00006479
4-Amino-TEMPO
Polymerization reagent
Packaging Glass Bottle
Quantity 250mg
Infrared Spectrum Authentic
Color Undesignated
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,chloroform,hexane
Formula Weight 171.26
Physical Form Adhering Crystals, Needles or Moist Mass
Percent Purity 97%
Chemical Name or Material 4-Amino-2,2,6,6-tetramethylpiperidinooxy
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 238-738-2