Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chloro-3-nitropyridine 97.0+%, TCI America™
SDP

Catalog No. A27981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27981G 1 g
A2798200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27981G Supplier TCI America Supplier No. A27981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2789-25-5
Molecular Formula C5H4ClN3O2
Molecular Weight (g/mol) 173.556
MDL Number MFCD07368898
InChI Key PDQAWJXOYURKPI-UHFFFAOYSA-N
PubChem CID 10965014
IUPAC Name 2-chloro-3-nitropyridin-4-amine
SMILES C1=CN=C(C(=C1N)[N+](=O)[O-])Cl

Specifications

Melting Point 187°C
Color Yellow
Quantity 1 g
Formula Weight 173.56
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloro-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.