Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-hydroxypyridine, 97%

Catalog No. p-4813056
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 38767-72-5 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38767-72-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
InChI Key SBQVQYPJWLJRQT-UHFFFAOYSA-N
PubChem CID 573530
IUPAC Name 4-amino-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1N

Specifications

Melting Point 206.5°C to 208.5°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-aminopyridin-2-ol, 4-amino-2-hydroxypyridine, 4-amino-pyridin-2-ol, 4-aminopyridin-2 1h-one, 4-amino-2-pyridone, 3-deazacytosine, 2 1h-pyridinone, 4-amino, 2 1h-pyridinone,4-amino, 2-pyridinol, 4-amino, 4-amino-1,2-dihydropyridin-2-one
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 4-Amino-2-hydroxypyridine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.