Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Amino-2-methylpyridine, 98%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.
Solubility
Soluble in methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.
Solubility
Soluble in methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 93°C to 97°C |
| Quantity | 1 g |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 108.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Amino-2-methylpyridine |
Safety and Handling
| Hazard Category | H302+H332-H315-H318-H335 |
| Hazard Statement | GHS H Statement H301-H318-H315-H335 Toxic if swallowed. Causes serious eye damage. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction