Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-methylpyridine, 98%

Catalog No. AAH5503606
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 18437-58-6 | C6H8N2 | 108.144 g/mol

4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18437-58-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00186509
InChI Key GNCLPNMQEGMNTG-UHFFFAOYSA-N
Synonym 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118
PubChem CID 728670
IUPAC Name 2-methylpyridin-4-amine
SMILES CC1=NC=CC(=C1)N

Specifications

Melting Point 93°C to 97°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 4-Amino-2-methylpyridine
Hazard Category H302+H332-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.