Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3,5-dichlorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. A190025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A190025G 25 g
A19005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A190025G Supplier TCI America Supplier No. A190025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24279-39-8
Molecular Formula C7H4Cl2F3N
Molecular Weight (g/mol) 230.01
MDL Number MFCD00052918
InChI Key ITNMAZSPBLRJLU-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl aniline, 4-amino-3,5-dichlorobenzotrifluoride, 3,5-dichloro-4-amino benzotrifluoride, 3,5-dichloro-4-aminobenzotrifluoride, 2,6-dichloro-4-trifluoromethyl benzenamine, benzenamine, 2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl phenylamine, 2,6-dichloro-4-trifluoromethyl-benzenamine, pubchem2187
PubChem CID 141094
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F

Specifications

Melting Point 37°C
Boiling Point 62°C
Quantity 25 g
UN Number 3077
Formula Weight 230.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3,5-dichlorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.