Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3,5-difluoropyridine, 97%

Catalog No. ac452530010
Click to view available options
Quantity:
1 g
5 g

CAS: 159783-22-9 | C5H4F2N2 | 130.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159783-22-9
Molecular Formula C5H4F2N2
Molecular Weight (g/mol) 130.1
InChI Key XWAOYJHUAQFFMT-UHFFFAOYSA-N
PubChem CID 2737654
IUPAC Name 3,5-difluoropyridin-4-amine
SMILES C1=C(C(=C(C=N1)F)N)F

Specifications

Melting Point 102°C to 105°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 4-amino-3,5-difluoropyridine, 3,5-difluoro-4-pyridinamine, 3,5-difluoro-pyridin-4-ylamine, 4-pyridinamine, 3,5-difluoro, difluoropyridinamine, 3,5-difluoro-4-pyridylamine, 3,5-difluoropyridin-4-ylamine, 4-pyridinamine,3,5-difluoro-9ci, 4-pyridinamine, 3,5-difluoro-9ci
Formula Weight 130.1
Percent Purity 97%
Chemical Name or Material 4-Amino-3, 5-difluoropyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.