Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%

Catalog No. AA3629014
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 116-63-2 | C10H9NO4S | 239.245 g/mol

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as a key starting material for the preparation of naphthalenesulfonic acid derivatives by reacting with palmitoyl chloride, which shows the activity to inhibit DNA polymerase and ribonuclease H Activities of HIV-1 reverse transcriptase. It is also used to prepare a copolyner with melamine, which is associated with graphene to prepare molecularly imprinted sensor and utilized for the determination of melatonin in human biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
MDL Number MFCD00004019
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
PubChem CID 8316
ChEBI CHEBI:19024
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

Specifications

Melting Point ∼295°C (decomposition)
Quantity 25 g
Beilstein 2697469
Merck Index 14,454
Solubility Information Soluble in hot water,hot hydrogen sulfite of sodium and alkali. Insoluble in water,ethanol,ethyl ether and benzene.
Formula Weight 239.25
Percent Purity ≥98%
Grade ACS
Physical Form Powder
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-147-3
RTECSNumber QK1292000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.