Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3-methylbenzoic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
2486-70-6
C8H9NO2
151.165
MFCD00007736
NHFKECPTBZZFBC-UHFFFAOYSA-N
3-methyl-4-aminobenzoic acid,4-amino-m-toluic acid,benzoic acid, 4-amino-3-methyl,4-amino-3-methyl benzoic acid,4-amino-3-methyl benzoate,4-amino-3-methylbenzoicacid,unii-201cxt4h5v,4-amino-3-methyl-benzoic acid,benzoic acid, 3-methyl-4-amino,4-amino-3-methylbenzenecarboxylic acid
75598
4-amino-3-methylbenzoic acid
CC1=C(C=CC(=C1)C(=O)O)N
2486-70-6
C8H9NO2
151.165
MFCD00007736
NHFKECPTBZZFBC-UHFFFAOYSA-N
3-methyl-4-aminobenzoic acid,4-amino-m-toluic acid,benzoic acid, 4-amino-3-methyl,4-amino-3-methyl benzoic acid,4-amino-3-methyl benzoate,4-amino-3-methylbenzoicacid,unii-201cxt4h5v,4-amino-3-methyl-benzoic acid,benzoic acid, 3-methyl-4-amino,4-amino-3-methylbenzenecarboxylic acid
75598
4-amino-3-methylbenzoic acid
CC1=C(C=CC(=C1)C(=O)O)N
Melting Point 169°C to 171°C
Quantity 5g
Beilstein 2802615
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 4-Amino-3-methylbenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-629-9

TSCA : No

Recommended Storage : Ambient temperatures