Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3-methylphenol, 98%

Click to view available options
Quantity:
100g
2835-99-6
C7H9NO
123.155
MFCD00007871
QGNGOGOOPUYKMC-UHFFFAOYSA-N
4-amino-m-cresol,4-hydroxy-2-methylaniline,3-methyl-4-aminophenol,p-amino-m-cresol,phenol, 4-amino-3-methyl,p-hydroxy-o-toluidine,2-methyl-4-hydroxyaniline,2-amino-5-hydroxytoluene,m-cresol, 4-amino,4-amino-3-cresol
17819
CHEBI:55546
4-amino-3-methylphenol
CC1=C(C=CC(=C1)O)N
2835-99-6
C7H9NO
123.155
MFCD00007871
QGNGOGOOPUYKMC-UHFFFAOYSA-N
4-amino-m-cresol,4-hydroxy-2-methylaniline,3-methyl-4-aminophenol,p-amino-m-cresol,phenol, 4-amino-3-methyl,p-hydroxy-o-toluidine,2-methyl-4-hydroxyaniline,2-amino-5-hydroxytoluene,m-cresol, 4-amino,4-amino-3-cresol
17819
CHEBI:55546
4-amino-3-methylphenol
CC1=C(C=CC(=C1)O)N
Quantity 100g
Melting Point 176°C to 179°C
UN Number UN3077
Beilstein 2078803
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-Amino-3-methylphenol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : GO6826000

TSCA : TSCA