Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-nitrobenzonitrile, 98%

Catalog No. p-7027025
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 6393-40-4 | C7H5N3O2 | 163.14 g/mol

4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

Solubility
Soluble in ethanol and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6393-40-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00013373
InChI Key JAHADAZIDZMHOP-UHFFFAOYSA-N
Synonym 3-nitro-4-aminobenzonitrile, 4-cyano-2-nitroaniline, benzonitrile, 4-amino-3-nitro, 4-amino-3-nitro-benzonitrile, 2-nitro-4-cyanoaniline, 4-amino-3-nitrobenzenecarbonitrile, zlchem 228, pubchem4669, 4-ammo-3-nitrobenzonitrile
PubChem CID 595901
IUPAC Name 4-amino-3-nitrobenzonitrile
SMILES NC1=CC=C(C=C1[N+]([O-])=O)C#N

Specifications

Melting Point 159°C to 161°C
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in ethanol and benzene.
Formula Weight 163.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitrobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.