Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-4'-chlorodiphenyl ether, 97%

Catalog No. p-7025549
Click to view available options
Quantity:
10 g
50 g

CAS: 101-79-1 | C12H10ClNO | 219.67 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-79-1
Molecular Formula C12H10ClNO
Molecular Weight (g/mol) 219.67
MDL Number MFCD00043925
InChI Key YTISFYMPVILQRL-UHFFFAOYSA-N
Synonym 4-4-chlorophenoxy aniline, 4-amino-4'-chlorodiphenyl ether, benzenamine, 4-4-chlorophenoxy, p-p-chlorophenoxy aniline, 4-4-chlorophenoxy benzenamine, 4-chloro-4'-aminodiphenyl ether, aniline, p-chlorophenoxy, 4-amino-4-chlorodiphenyl ether, 4'-chloro-4-aminobiphenyl ether, aniline, p-p-chlorophenoxy
PubChem CID 7578
IUPAC Name 4-(4-chlorophenoxy)aniline
SMILES NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 100°C to 103°C
Odor Odorless
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 219.67
Percent Purity 97%
Chemical Name or Material 4-Amino-4'-chlorodiphenyl ether
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 202-976-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.