Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. A29181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29181G 1 g
A2918200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29181G Supplier TCI America Supplier No. A29181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123654-26-2
Molecular Formula C9H8ClNO3
Molecular Weight (g/mol) 213.617
MDL Number MFCD12923204
InChI Key KRMUVKSAOVLXLF-UHFFFAOYSA-N
Synonym 4-Amino-5-chlorocoumaran-7-carboxylic Acid
PubChem CID 10632401
IUPAC Name 4-amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid
SMILES C1COC2=C1C(=C(C=C2C(=O)O)Cl)N

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 1 g
Formula Weight 213.62
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.