Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-chloro-2-ethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A194125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A194125G 25 g
A19415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A194125G Supplier TCI America Supplier No. A194125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108282-38-8
Molecular Formula C9H10ClNO3
Molecular Weight (g/mol) 215.63
MDL Number MFCD03840527
InChI Key XWGYOMHQGQZRLC-UHFFFAOYSA-N
PubChem CID 10536658
IUPAC Name 4-amino-5-chloro-2-ethoxybenzoic acid
SMILES CCOC1=CC(N)=C(Cl)C=C1C(O)=O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-chloro-2-ethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.