Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-L-phenylalanine, 95%

Catalog No. p-7034299
Click to view available options
Quantity:
1 g
5 g

CAS: 943-80-6 | C9H12N2O2 | 180.21 g/mol

It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943-80-6
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00069927
InChI Key CMUHFUGDYMFHEI-QMMMGPOBSA-N
Synonym 4-amino-l-phenylalanine, h-phe 4-nh2-oh, 4-aminophenylalanine, l-4-aminophenylalanine, p-amino-l-phenylalanine, p-aminophenylalanine, s-2-amino-3-4-aminophenyl propanoic acid, 4-amino-phenylalanine, 2s-2-amino-3-4-aminophenyl propanoic acid, l-4-nh2-phe-oh
PubChem CID 151001
ChEBI CHEBI:29737
IUPAC Name (2S)-2-amino-3-(4-aminophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Specifications

Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 180.2
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material 4-Amino-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.