Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoantipyrine, 97%

Catalog No. aaa1384622
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 83-07-8 | C11H13N3O | 203.245 g/mol

4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

Solubility
Soluble in water. 500 g/L (20°C), methanol. Partially soluble in diethyl ether.

Notes
Incompatible with Strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-07-8
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD00003145
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
PubChem CID 2151
ChEBI CHEBI:59026
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N

Specifications

Melting Point 106°C to 110°C
Assay Percent Range 97%
Quantity 100 g
Beilstein 181635
Merck Index 14,591
Solubility Information Soluble in water. 500g/L (20°C),methanol. Partially soluble in diethyl ether.
Formula Weight 203.25
Percent Purity 97%
Chemical Name or Material 4-Aminoantipyrine
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-452-3
RTECSNumber CD2480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.