Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoantipyrine, 99%, MP Biomedicals™

Catalog No. p-4404651
Click to view available options
Quantity:
10 g
100 g
250 g

4-Aminoantipyrine, Yellow Powder

  • Reagent for determination of chlorinated phenols, phenylglycosidase, and glucose in the presence of phenol and peroxidase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H13N3O
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
PubChem CID 2151

Specifications

Color Yellow
Quantity 10 g
Synonym 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone
Molecular Weight (g/mol) 203.25
Physical Form Powder
RTECSNumber CD2480000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.