Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
2835-68-9
C7H8N2O
136.15
MFCD00007999
QIKYZXDTTPVVAC-UHFFFAOYSA-N
p-aminobenzamide,benzamide, 4-amino,4-amino-benzamide,p-carbamoylaniline,p-aminobenzoic acid amide,benzamide, p-amino,4-amino benzamide,unii-77722i6pac,ccris 6792,aminobenzamide-4
76079
4-aminobenzamide
C1=CC(=CC=C1C(=O)N)N
2835-68-9
C7H8N2O
136.15
MFCD00007999
QIKYZXDTTPVVAC-UHFFFAOYSA-N
p-aminobenzamide,benzamide, 4-amino,4-amino-benzamide,p-carbamoylaniline,p-aminobenzoic acid amide,benzamide, p-amino,4-amino benzamide,unii-77722i6pac,ccris 6792,aminobenzamide-4
76079
4-aminobenzamide
C1=CC(=CC=C1C(=O)N)N
Melting Point 181.0°C to 185.0°C
Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H4CONH2
Solubility Information Solubility in water: soluble in water.,soluble in ethanol
Formula Weight 136.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Aminobenzamide
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-612-3