Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzoic Acid, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
150-13-0
C7H7NO2
137.14
MFCD00007894
ALYNCZNDIQEVRV-UHFFFAOYSA-N
p-aminobenzoic acid,paba,para-aminobenzoic acid,vitamin bx,sunbrella,p-carboxyaniline,4-carboxyaniline,hachemina,paraminol,benzoic acid, 4-amino
978
CHEBI:30753
4-aminobenzoic acid
C1=CC(=CC=C1C(=O)O)N
150-13-0
C7H7NO2
137.14
MFCD00007894
ALYNCZNDIQEVRV-UHFFFAOYSA-N
p-aminobenzoic acid,paba,para-aminobenzoic acid,vitamin bx,sunbrella,p-carboxyaniline,4-carboxyaniline,hachemina,paraminol,benzoic acid, 4-amino
978
CHEBI:30753
4-aminobenzoic acid
C1=CC(=CC=C1C(=O)O)N
Infrared Spectrum Authentic
Melting Point 186.0°C to 189.0°C
Quantity 1kg
Color White to Yellow
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2NC6H4CO2H
Beilstein 14, 418
Fieser 02,24
Merck Index 15, 418
Solubility Information Solubility in water: 4.7g/L (20°C). Other solubilities: soluble in glacial acetic acid,soluble in ethyl acetate,slightly soluble in benzene,121g/L etanol- 1g/50mL ether,soluble in chloroform
Formula Weight 137.14
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Aminobenzoic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning