Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzonitrile, 98%

Catalog No. AC164470500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164470500 50 g Glass bottle
AC164470100 10 g Glass bottle
AC164472500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164470500 Supplier Thermo Scientific Chemicals Supplier No. 164470500
Add to Cart

CAS: 873-74-5 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-74-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007821
InChI Key YBAZINRZQSAIAY-UHFFFAOYSA-N
Synonym 4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
PubChem CID 13396
IUPAC Name 4-aminobenzonitrile
SMILES C1=CC(=CC=C1C#N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Color Beige to Orange-Yellow or White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CN
Quantity 50 g
Beilstein 14, 425
Solubility Information Solubility in water: insoluble
Formula Weight 118.14
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Aminobenzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective glo
EINECSNumber 212-850-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.