Learn More
Description
- Pyridin-3-yl sulfurofluoridate can be used in the pd-catalyzed suzuki-miyuara cross-coupling reaction
- The fluorosulfate functionality was shown by sharpless and coworkers to have distinct reactivity in comparison to other functional groups used in suzuki-miyuara reactions, such as bromides, chlorides and triflates
- This inherent property provides a strong potential for selective suzuki-miyuara couplings for chemoselective reaction sequences.palladium-catalyzed; ligand-free suzuki reaction in water using aryl fluorosulfates chemoselective synthesis of polysubstituted pyridines from heteroaryl fluorosulfates
Specifications
Specifications
| CAS | 3544-25-0 |
| Melting Point | 45°C to 48°C (lit.) |
| Boiling Point | 312°C (lit.) |
| Molecular Formula | C8H8N2 |
| Linear Formula | H2NC6H4CH2CN |
| MDL Number | MFCD00007912 |
| Quantity | 500 mg |
| Synonym | 4-Aminophenylacetonitrile |
| Molecular Weight (g/mol) | 132.16 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.