Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Aminobiphenyl, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used to manufacture azo dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to manufacture azo dyes.
Solubility
Soluble in Dichloromethane, DMSO and Methanol
Notes
Store at -20°C. Avoid heat and oxidizing agents.
It is used to manufacture azo dyes.
Solubility
Soluble in Dichloromethane, DMSO and Methanol
Notes
Store at -20°C. Avoid heat and oxidizing agents.

Specifications
Specifications
Melting Point | 52°C to 54°C |
Boiling Point | 191°C (15 mmHg) |
Quantity | 5 g |
UN Number | UN3077 |
Beilstein | 386533 |
Merck Index | 14,1235 |
Solubility Information | Soluble in Dichloromethane,DMSO and Methanol |
Formula Weight | 169.23 |
Percent Purity | 98% |
Chemical Name or Material | 4-Aminobiphenyl |
Safety and Handling
Hazard Category | H302-H350 |
Hazard Statement | GHS H Statement H350-H302 May cause cancer. Harmful if swallowed. |
Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 202-177-1 |
RTECSNumber | DU8925000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction