Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 4-Aminobiphenyl
Click to view available options
Quantity:
1 g
Description
- Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
- Sigma-aldrich does not collect analytical data for this product
- Buyer assumes responsibility to confirm product identity and/or purity
- All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise
Specifications
Specifications
| CAS | 92-67-1 |
| Melting Point | 52°C to 54°C (lit.) |
| Boiling Point | 191°C (15 mmHg) |
| Molecular Formula | C12H11N |
| Linear Formula | C6H5C6H4NH2 |
| MDL Number | MFCD00007879 |
| Quantity | 1 g |
| Synonym | 4-Aminodiphenyl; 4-Biphenylamine; 4-Biphenylylamine; 4-Phenylaniline; Biphenyl-4-ylamine; NSC 7660; Xenylamine |
| Molecular Weight (g/mol) | 169.22 |
| Percent Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction