Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 4-aminobiphenyl DNA Monoclonal Antibody (4C11)
GREENER_CHOICE

Catalog No. AMPA6590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6590 Supplier Invitrogen™ Supplier No. MA116590
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

4-aminobiphenyl DNA Monoclonal Antibody for ICC/IF, IHC, ELISA

4-aminoBiphenyl (4-ABP) is an aromatic amine (arylamine). 4-ABP can be found as a constituent of tobacco smoke, in the form of an environmental carcinogen. It is metabolized by hepatic cytochrome P4501A2 to yield a direct-acting mutagen known as N-hydroxy-aminobiphenyl. Yet, the accumulation of biologically active metabolites is dependent on the activation and detoxification of N-acetylation. 4-ABP has been shown to be a bladder carcinogen in humans and rodents. As well as genetic factors, 4-ABP may also play a role in breast cancer. It is hypothesized that smokers may have higher levels of DNA adducts than non-smokers and that the role of 4-ABP-DNA adducts and their relationship to tobacco exposure may be related to breast tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-aminobiphenyl DNA
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 4C11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene Alias 4-AMBP DNA; 4-aminobiphenyl DNA
Host Species Mouse
Immunogen 4-amino biphenyl DNA.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.