Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobutyraldehyde diethyl acetal, 95%, Thermo Scientific™

Catalog No. AAL1428614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1428614 25 g
AAL1428606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1428614 Supplier Thermo Scientific Chemicals Supplier No. L1428614

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Aminobutyraldehyde diethyl acetal is used as a pharmaceutical intermediate.

Solubility
Miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agent, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6346-09-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.25
MDL Number MFCD00008227
InChI Key GFLPSABXBDCMCN-UHFFFAOYSA-N
Synonym 4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine #
PubChem CID 80672
IUPAC Name 4,4-diethoxybutan-1-amine
SMILES CCOC(CCCN)OCC

Specifications

Boiling Point 196°C to 198°C
Flash Point 63°C (145°F)
Quantity 25 g
UN Number UN2735
Beilstein 1739649
Formula Weight 161.25
Percent Purity 95%
Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.