Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobutyric Acid, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600136
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

AM151, 56-12-2, C4H9NO2

Spectrum™ Chemical 4-Aminobutyric Acid, Reagent belongs to the transaminases family which transfer nitrogenous groups. It is an enzyme which plays a part in 5 metabolic pathways: glutamate metabolism, beta-alanine metabolism, alanine and aspartate metabolism, butanoate metabolism, and propionate metabolism. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
IUPAC Name 4-aminobutanoic acid
SMILES NCCCC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.33%
Loss on Drying 0.5%
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.