Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Aminomethyl)-2-methoxyphenol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A233025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A233025G 25 g
A23305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A233025G Supplier TCI America Supplier No. A233025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10-2-7149
Molecular Formula C8H11ClNO2
Molecular Weight (g/mol) 188.63
MDL Number MFCD00012864
InChI Key LOYPVODLNGWOLM-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methoxybenzylamine hydrochloride, 4-aminomethyl-2-methoxyphenol hydrochloride, vanillylamine hydrochloride, n-vanillylamine hydrochloride, 2-vanillylamine hydrochloride, 4-hydroxy-3-methoxybenzylamine hcl, phenol, 4-aminomethyl-2-methoxy-, hydrochloride, 4-aminomethyl-2-methoxyphenol, chloride, vanillylamine hcl
PubChem CID 165576
IUPAC Name 4-(aminomethyl)-2-methoxyphenol;hydrochloride
SMILES COC1=C(C=CC(=C1)CN)O.Cl

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 25 g
Formula Weight 189.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Aminomethyl)-2-methoxyphenol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.