Learn More
4-(Aminomethyl)indole, 97%
Description
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 130°C to 135°C |
Quantity | 1 g |
Solubility Information | Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. |
Formula Weight | 146.19 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Aminomethyl)indole |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.