Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Aminomethyl)pyridine, 98%

Catalog No. p-4813003
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 3731-53-1 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-53-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
InChI Key TXQWFIVRZNOPCK-UHFFFAOYSA-N
Synonym 4-pyridinemethaneamine, 4-aminomethyl pyridine, 4-pyridinemethanamine, 4-pyridylmethylamine, 4-picolylamine, 4-aminomethylpyridine, 4-picolinylamine, 4-pyridinemethylamine, 1-pyridin-4-ylmethanamine, 4-picolinamine
PubChem CID 77317
IUPAC Name pyridin-4-ylmethanamine
SMILES C1=CN=CC=C1CN

Specifications

Melting Point -8.0°C
Density 1.0650g/mL
Boiling Point 228.0°C to 230.0°C
Flash Point 108°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5515
Quantity 25 g
Specific Gravity 1.065
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 4-(Aminomethyl)pyridine
Show More Show Less
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 223-092-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.