Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenol, 98%

Catalog No. p-7021300
Click to view available options
Quantity:
50 g
250 g
1000 g
5000 g

Catalog No. AAA1358118

Supplier: Thermo Scientific Chemicals A1358118

Only null left
Add to Cart

CAS: 123-30-8 | C6H7NO | 109.128 g/mol

4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

Solubility
Soluble in acetonitrile, ethyl acetate, acetone, hot water, dimethylsulfoxide and alkalies. Slightly soluble in toluene, diethyl ether, ethanol, trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.

Notes
Incompatible with acids, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-30-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007869
InChI Key PLIKAWJENQZMHA-UHFFFAOYSA-N
Synonym p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PubChem CID 403
ChEBI CHEBI:17602
IUPAC Name 4-aminophenol
SMILES C1=CC(=CC=C1N)O

Specifications

Melting Point 186°C to 190°C
Density 1.29
Boiling Point 284°C (decomposition)
Flash Point 195°C (383°F)
Odor Phenol-like
Assay Percent Range 98%
Quantity 50 g
UN Number UN2512
Beilstein 385836
Sensitivity Air and light sensitive
Merck Index 14,462
Solubility Information Soluble in acetonitrile,ethyl acetate,acetone,hot water,dimethylsulfoxide and alkalies. Slightly soluble in toluene,diethyl ether,ethanol,trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 4-Aminophenol
Show More Show Less
Hazard Category H302+H332-H341
Hazard Statement GHS H Statement
H341-H302-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280-P301+P312-P304+P340-P308+P313-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-)
EINECSNumber 204-616-2
RTECSNumber SJ5075000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.