Learn More
4-Aminosalicylic acid, 98+%
Description
Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.
Solubility
Soluble in water (2 g/L) at 20°C, dilute sodium hydroxide, dilute nitric acid, ether (slightly), methanol, acetone, and ethanol. Virtually insoluble in benzene, chloroform, and carbon tetrachloride.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
            Specifications
Specifications
| Melting Point | ∼135°C to 145°C (decomposition) | 
| Odor | Odorless | 
| Quantity | 100 g | 
| Beilstein | 473071 | 
| Merck Index | 14,477 | 
| Solubility Information | Soluble in water (2g/L) at 20°C,dilute sodium hydroxide,dilute nitric acid,ether (slightly),methanol,acetone,and ethanol. Virtually insoluble in benzene,chloroform,and carbon tetrachloride. | 
| Formula Weight | 153.14 | 
| Percent Purity | ≥98% | 
| Chemical Name or Material | 4-Aminosalicylic acid | 
Safety and Handling
| Hazard Category | H315-H319-H335 | 
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.  | 
                                
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c | 
| EINECSNumber | 200-613-5 | 
| RTECSNumber | VO1225000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.