Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminostilbene 97.0+%, TCI America™
SDP

Catalog No. A085425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A085425G 25 g
A08545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A085425G Supplier TCI America Supplier No. A085425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 834-24-2
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00025377
InChI Key VFPLSXYJYAKZCT-VOTSOKGWSA-N
Synonym 4-Stilbenamine
PubChem CID 1272788
IUPAC Name 4-[(1E)-2-phenylethenyl]aniline
SMILES NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1

Specifications

Melting Point 151°C
Color Yellow
Quantity 25 g
Formula Weight 195.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminostilbene
RTECSNumber WJ3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.