Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzoylbiphenyl, 99%

Catalog No. p-7022685
Click to view available options
Quantity:
25 g
100 g

CAS: 2128-93-0 | C19H14O | 258.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2128-93-0
Molecular Formula C19H14O
Molecular Weight (g/mol) 258.32
MDL Number MFCD00003079
InChI Key LYXOWKPVTCPORE-UHFFFAOYSA-N
Synonym 4-benzoylbiphenyl, 4-phenylbenzophenone, p-phenylbenzophenone, benzophenone, 4-phenyl, p-benzoylbiphenyl, 4-diphenyl phenyl ketone, p-biphenylyl phenyl ketone, trigonal 12, 4-biphenylyl phenyl ketone, methanone, 1,1'-biphenyl-4-ylphenyl
PubChem CID 75040
IUPAC Name phenyl-(4-phenylphenyl)methanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 98°C to 103°C
Quantity 25 g
Beilstein 1876092
Formula Weight 258.32
Percent Purity 99%
Chemical Name or Material 4-Benzoylbiphenyl
EINECSNumber 218-345-2
RTECSNumber PC4936800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.