Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%

Catalog No. AC155000025
Click to view available options
Quantity:
25 g
500 g
2.5 kg

trans-1,2-Diaminocyclohexane-N,N,N,N-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4 | CAS: 125572-95-4 | C14H20N2O8 | 344.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
MDL Number MFCD00149243,MFCD00066429,MFCD00003845
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
Synonym 2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
PubChem CID 2723844
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

Melting Point >210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula C6H10[N(CH2CO2H)2]2·H2O
Quantity 2.5 kg
Beilstein 13,III,1
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in most common organic solvents, soluble in alkali solutions
IUPAC Name 2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
Formula Weight 364.35
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.