Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyphenylacetyl Chloride 96.0+%, TCI America™
SDP

Catalog No. B17805G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 39188-62-0
Molecular Formula C15H13ClO2
Molecular Weight (g/mol) 260.717
MDL Number MFCD00674192
InChI Key GZAGJMNAVUCWGM-UHFFFAOYSA-N
PubChem CID 11357442
IUPAC Name 2-(4-phenylmethoxyphenyl)acetyl chloride
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)Cl

Specifications

Melting Point 78°C
Quantity 5 g
UN Number 3261
Formula Weight 260.72
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyphenylacetyl Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.