Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylphenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AC433920010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433920010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433920010 Supplier Thermo Scientific Chemicals Supplier No. 433920010

Chemical Identifiers

CAS 911708-01-5
Molecular Formula C19H23BO2
Molecular Weight (g/mol) 294.20
MDL Number MFCD09266180
InChI Key FPLGWDLJBAAWHO-UHFFFAOYSA-N
Synonym 4-benzylphenylboronic acid pinacol ester, 2-4-benzylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb035, 4-benzylbenzeneboronic acid pinacol ester
PubChem CID 46739038
IUPAC Name 2-(4-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 93.8°C to 95°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 294.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzylphenylboronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.