Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylthiomorpholine 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. B243425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B243425G 25 g
B24345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B243425G Supplier TCI America Supplier No. B243425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26475-66-1
Molecular Formula C11H15NO2S
Molecular Weight (g/mol) 225.31
MDL Number MFCD00084894
InChI Key KFAMTQFKYUXQKV-UHFFFAOYSA-N
PubChem CID 318454
IUPAC Name 4-benzyl-1λ⁶-thiomorpholine-1,1-dione
SMILES O=S1(=O)CCN(CC2=CC=CC=C2)CC1

Specifications

Color White
Quantity 25 g
Synonym 4-benzylthiomorpholine 1,1-dioxide, 1,1-dioxide-4-phenylmethyl thiomorpholine, 4-benzyl-1??-thiomorpholine-1,1-dione, 4-benzylthiomorpholine1,1-dioxide, acmc-209grq, maybridge1_003205, 4-benzylthiomorpholine-1,1-dioxide, 4-benzylthiomorpholine-1,1,-dioxide, 4-benzyl-1,4-thiazaperhydroine-1,1-dione
Formula Weight 225.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzylthiomorpholine 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.