Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenyl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B186625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2170-13-0
Molecular Formula C19H14O2
Molecular Weight (g/mol) 274.319
MDL Number MFCD00223541
InChI Key CINHWMYRCOGYIX-UHFFFAOYSA-N
Synonym Benzoic Acid 4-Biphenyl Ester
PubChem CID 231743
IUPAC Name (4-phenylphenyl) benzoate
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)OC(=O)C3=CC=CC=C3

Specifications

Melting Point 151°C
Color White
Quantity 25 g
Formula Weight 274.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenyl Benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.