Learn More
4-Bromo-1-butanol, tech., cont. varying amounts of THF
Description
4-Bromo-1-butanol is used to prepare 2-(4-bromo-butoxy)-tetrahydro-pyran by reacting with 3,4-dihydro-2H-pyran in the presence of amberlyst 15 as a catalyst. It reacts with dihydropyran to prepare tetrahydropyranyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow |
Boiling Point | 63°C to 65°C (5 mmHg) |
Flash Point | 32°C (91°F) |
Refractive Index | 1.5035 |
Linear Formula | Br(CH2)4OH |
Quantity | 5 g |
UN Number | UN1987 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Immiscible with water. |
Formula Weight | 153.02 |
Show More |
Safety and Handling
Hazard Category | H225-H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H226-H351-H315-H319 Flammable liquid and vapor. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 3; Packaging Group: II |
EINECSNumber | 251-355-5 |
TSCA | No |
storageNote1 | −30°C to −10°C |
Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.