Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-butene, 97%

Catalog No. p-7081144
Click to view available options
Quantity:
5 g
25 g

CAS: 5162-44-7 | C4H7Br | 135.004 g/mol

4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.

Solubility
Miscible with water, benzene, ether and ethanol.

Notes
Light sensitive. Store in a cool place. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5162-44-7
Molecular Formula C4H7Br
MDL Number MFCD00000258
InChI Key DMAYBPBPEUFIHJ-UHFFFAOYSA-N
Synonym 4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
PubChem CID 21241
IUPAC Name 4-bromobut-1-ene

Specifications

Density 1.33
Boiling Point 98°C to 100°C
Flash Point 1°C (34°F)
Refractive Index 1.463
Quantity 5 g
UN Number UN1993
Beilstein 1098377
Sensitivity Light sensitive
Solubility Information Miscible with water,benzene,ether and ethanol.
SMILES C=CCCBr
Molecular Weight (g/mol) 135.004
Formula Weight 135
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-butene
Show More Show Less
Hazard Category H225-H317-H319-H334-H335
Hazard Statement GHS H Statement
H225-H334-H335-H319-H317
Highly flammable liquid and vapor.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P333+P313-P342+P311-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 225-937-4
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.