Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Bromo-1-butyne, 97%
Click to view available options
Quantity:
5 g
25 g
Description
4-Bromo-1-butyne is used for pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-1-butyne is used for pharmaceutical intermediates.
Solubility
Immiscible with water.
Notes
Recommended storage in refrigerator. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
4-Bromo-1-butyne is used for pharmaceutical intermediates.
Solubility
Immiscible with water.
Notes
Recommended storage in refrigerator. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 1.417 |
| Boiling Point | 110°C |
| Flash Point | 24°C (75°F) |
| Refractive Index | 1.481 |
| Linear Formula | HC≡CCH2CH2Br |
| Quantity | 25 g |
| UN Number | UN1992 |
| Solubility Information | Immiscible with water. |
| Formula Weight | 132.99 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H226-H301-H317 |
| Hazard Statement | GHS H Statement H301-H226-H317 Toxic if swallowed. Flammable liquid and vapor. May cause an allergic skin reaction. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P330-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction