Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-chloro-2-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. B437425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B437425G 25 g
B43745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B437425G Supplier TCI America Supplier No. B437425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16588-24-2
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.45
MDL Number MFCD05863216
InChI Key DJRYWPGOQTUJMQ-UHFFFAOYSA-N
Synonym 5-Bromo-2-chloronitrobenzene
PubChem CID 817974
IUPAC Name 4-bromo-1-chloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=C(Cl)C=CC(Br)=C1

Specifications

Melting Point 72°C
Color Yellow
Quantity 25 g
Formula Weight 236.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1-chloro-2-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.