Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-bromo-1-fluoro-2-nitrobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
364-73-8
C6H3BrFNO2
220
MFCD00129165
UQEANKGXXSENNF-UHFFFAOYSA-N
5-bromo-2-fluoronitrobenzene,1-bromo-4-fluoro-3-nitrobenzene,2-fluoro-5-bromonitrobenzene,4-bromo-1-fluoro-2-nitro-benzene,benzene, 4-bromo-1-fluoro-2-nitro,pubchem1046,5-bromo-fluoronitrobenzene,acmc-1cpc6,5-bromo-2-fluronitrobenzene,ksc226i7f
2736328
4-bromo-1-fluoro-2-nitrobenzene
C1=CC(=C(C=C1Br)[N+](=O)[O-])F
364-73-8
C6H3BrFNO2
220
MFCD00129165
UQEANKGXXSENNF-UHFFFAOYSA-N
5-bromo-2-fluoronitrobenzene,1-bromo-4-fluoro-3-nitrobenzene,2-fluoro-5-bromonitrobenzene,4-bromo-1-fluoro-2-nitro-benzene,benzene, 4-bromo-1-fluoro-2-nitro,pubchem1046,5-bromo-fluoronitrobenzene,acmc-1cpc6,5-bromo-2-fluronitrobenzene,ksc226i7f
2736328
4-bromo-1-fluoro-2-nitrobenzene
C1=CC(=C(C=C1Br)[N+](=O)[O-])F
Refractive Index 1.5750 to 1.578
Melting Point 18°C to 20°C
Boiling Point 240°C to 241°C
Flash Point 110°C
Specific Gravity 1.786
Density 1.7860g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Brown to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H3(F)NO2
Formula Weight 220
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 4-Bromo-1-fluoro-2-nitrobenzene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning