Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific™

Catalog No. AAH6659003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6659003 1 g
AAH66590MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6659003 Supplier Thermo Scientific Chemicals Supplier No. H6659003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288246-16-2
Molecular Formula C4H2BrN3
Molecular Weight (g/mol) 171.985
MDL Number MFCD00105683
InChI Key ZXVFKQRZKKGVNJ-UHFFFAOYSA-N
Synonym 4-bromo-1h-pyrazole-3-carbonitrile, 4-bromo-2h-pyrazole-3-carbonitrile, 4-bromopyrazole-3-carbonitrile, 4-bromo-3-cyano-1h-pyrazole, 1h-pyrazole-3-carbonitrile, 4-bromo, pubchem10182, acmc-20aa8h, maybridge1_006470, ksc497k2j, buttpark 181\04-37
PubChem CID 2735623
IUPAC Name 4-bromo-1H-pyrazole-5-carbonitrile
SMILES C1=NNC(=C1Br)C#N

Specifications

Melting Point 155°C to 159°C
Color White to Yellow
Quantity 1 g
UN Number UN3439
Formula Weight 171.98
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-1H-pyrazole-3-carbonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.